Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant GST Epitope Tag Protein
SDP

Catalog No. NBC118537 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118537 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118537 Supplier Novus Biologicals™ Supplier No. NBC118537
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

An un-tagged recombinant protein corresponding to the amino acids 1-218 of Schistosoma japonicum E.coli (Bioactivity Validated) GST Epitope Tag The Recombinant Parasite GST Epitope Tag Protein is derived from E. coli. The Recombinant Parasite GST Epitope Tag Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

Accession Number NP_ P08515
For Use With (Application) Bioactivity, SDS-PAGE
Formulation PBS (pH 7.4)
Gene ID (Entrez) 2944
Molecular Weight (g/mol) 26kDa
Name GST Epitope Tag Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Glutathione S-Transferase(GST), 1-224aa, Schistosoma japonicum Recombinant, E.coli (NP_ P08515).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Non-species Specific
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.