Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 266743 |
| Species | Mouse |
| Purity | >70% |
| Content And Storage | Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.) |
| Formulation | PBS, BSA 0,1 % |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-06574. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | BHLHE79, bHLHe79neuronal PAS domain-containing protein 4, Class E basic helix-loop-helix protein 79, Le-PAS, neuronal PAS domain protein 4, Neuronal PAS4, NXFHLH-PAS transcription factor NXF, PASD10PAS domain-containing protein 10 |
| Gene Symbol | NPAS4 |
| Molecular Weight (g/mol) | M.W. (theoretical): 3.63 kDa |
| Product Type | Npas4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
