Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ NTF2 Protein

Click to view available options
Quantity:
0.1 mg
0.5 mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-127 of Human NTF2 The Recombinant Human NTF2 Protein is derived from E. coli. The Recombinant Human NTF2 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Concentration | 1.0mg/mL |
For Use With (Application) | ELISA, SDS-PAGE |
Formulation | In 20mM Tris-HCl Buffer (pH 8.0) containing 10% Glycerol |
Gene ID (Entrez) | 10204 |
Molecular Weight (g/mol) | 16.6kDa |
Purification Method | Protein |
Quantity | 0.1 mg |
Source | Human |
Immunogen | MGSSHHHHHH SSGLVPRGSH MGDKPIWEQI GSSFIQHYYQ LFDNDRTQLG AIYIDASCLT WEGQQFQGKA AIVEKLSSLP FQKIQHSITA QDHQPTPDSC IISMVVGQLK ADEDPIMGFH QMFLLKNIND AWVCTNDMFR LALHNFG |
Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction