Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PC-12 Whole Cell Lysate
SDP

Catalog No. NBP213170 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP213170 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP213170 Supplier R&D Systems™ Supplier No. NBP213170
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

The PC-12 Whole Cell Lysate is derived from rat. It has been validated for the following applications: Western Blot, Immunoprecipitation.

Specifications

Quantity 0.25 mg
Storage Buffer Standard RIPA buffer including protease and phosphotase inhibitors and 0.1% SDS
Test Specificity PC-12 (rat adrenal gland pheochromocytoma) whole cell lysate (non-denatured)
Cellular Fraction Whole
Content And Storage -80°C. Avoid freeze-thaw cycles.
Product Line Novus
Protein State Native
For Use With (Application) Immunoprecipitation, Western Blot
Tissue Nervous

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.