Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ PDIA6 Protein
SDP

Catalog No. NBP15085901 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15085901 0.1 mg
NBP1508595 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15085901 Supplier Novus Biologicals™ Supplier No. NBP1508590.1MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 20-440 of Human PDIA6 The Recombinant Human PDIA6 Protein is derived from E. coli. The Recombinant Human PDIA6 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 2mM DTT, 50mM NaCl
Gene ID (Entrez) 10130
Molecular Weight (g/mol) 48.5kDa
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. coli
Immunogen PDIA6, 20-440aa, Human, His tag, E. coli
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name PDIA6
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.