Learn More
Specifications
Specifications
| Gene ID (Entrez) | 5166 |
| Species | Human |
| Accession Number | Q16654 |
| Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
| Formulation | Peptide dissolved in dH2O. Contains no BSA. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NBP1-07047. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 4, mitochondrial, EC 2.7.11, EC 2.7.11.2, FLJ40832, Pyruvate dehydrogenase kinase isoform 4, pyruvate dehydrogenase kinase, isoenzyme 4, pyruvate dehydrogenase kinase, isozyme 4, pyruvate dehydrogenase, lipoamide, kinase isozyme 4, mitochondrial |
| Gene Symbol | PDK4 |
| Product Type | PDK4 |
| Quantity | 0.05 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
