Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ PGD2 Synthase/PTGDS Protein
SDP

Catalog No. NBP15107805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15107805 0.05 mg
NBP15107802 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15107805 Supplier Novus Biologicals™ Supplier No. NBP1510780.05MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 23-190 of Human PGD2 Synthase/PTGDS The Recombinant Human PGD2 Synthase/PTGDS Protein is derived from E. coli. The Recombinant Human PGD2 Synthase/PTGDS Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_000945
Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 30% glycerol, 1mM EDTA, 0.1M NaCl
Gene ID (Entrez) 5730
Molecular Weight (g/mol) 20.9kDa
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. coli
Immunogen Prostaglandin D Synthase/Lipocalin Protein, 23-190aa, Human, His tag, E. coli (NP_000945).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name PGD2 Synthase/PTGDS
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.