Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ PGD2 Synthase/PTGDS Protein
SDP

Catalog No. p-5355153
Click to view available options
Quantity:
0.05 mg
0.25 mg

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 23-190 of Human PGD2 Synthase/PTGDS The Recombinant Human PGD2 Synthase/PTGDS Protein is derived from E. coli. The Recombinant Human PGD2 Synthase/PTGDS Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_000945
Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 30% glycerol, 1mM EDTA, 0.1M NaCl
Gene ID (Entrez) 5730
Molecular Weight (g/mol) 20.9kDa
Purification Method SDS-PAGE
Quantity 0.05 mg
Source E. coli
Immunogen Prostaglandin D Synthase/Lipocalin Protein, 23-190aa, Human, His tag, E. coli (NP_000945).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name PGD2 Synthase/PTGDS
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.