Learn More
Description
Specifications
Specifications
| Concentration | Lyoph |
| For Use With (Application) | Bioactivity, SDS-PAGE |
| Formulation | Lyophilized from 50 mM Tris-HCl (pH 8.0), 2 mM Calcium acetate. |
| Molecular Weight (g/mol) | M.W. (theoretical): 28.9 kDa |
| Name | Proteinase K Protein |
| Purification Method | SDS-PAGE |
| Quantity | 500 mg |
| Storage Requirements | Store at -20 to -80 C. Avoid freeze-thaw cycles. |
| Reconstitution | Reconstitute with deionized H2O. |
| Purity or Quality Grade | >90%, by SDS-PAGE |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
