Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 138428 |
| Name | PTRH1 Protein |
| Purification Method | SDS-PAGE |
| Immunogen | The sequence of the first five N-terminal amino acids was determined and was found to be Ser-Val-Ser-Glu-Ile. The sequence of the first five N-terminal amino acids was determined and was found to be Ser-Val-Ser-Glu-Ile. |
| Storage Requirements | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
