Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ PUMA Antibody Blocking Peptide
SDP

Catalog No. NB500261PEP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500261PEP 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500261PEP Supplier Novus Biologicals™ Supplier No. NB500261PEP
Add to Cart
Add to Cart
A peptide to BBC3.

Specifications

Gene ID (Entrez) 27113
Species Human
Content And Storage Store at -80 C. Avoid freeze-thaw cycles.
Formulation Peptide dissolved in dH2O. Contains no BSA.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB500-261. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias BCL2 binding component 3, bcl-2-binding component 3, JFY1, JFY-1, p53 up-regulated modulator of apoptosis, PUMAFLJ42994
Gene Symbol BBC3
Product Type PUMA
Quantity 0.05 mg
Regulatory Status RUO - research use only
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.