Learn More
Novus Biologicals™ Raw 264.7 Whole Cell Lysate

Description
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Blood Lymph |
| Quantity | 0.1 mL |
| Storage Buffer | Standard RIPA buffer including protease and phosphatase inhibitors and 0.1% SDS diluted in 1X lithium dodecyl sulfate (LDS) sample buffer with bromophenol blue and 40-70% glycerol, pH 8.4. BME has not been added. |
| Cellular Fraction | Whole |
| Host Species | Mouse |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Denatured |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.