Learn More
Description
Specifications
Specifications
| Accession Number | NP_060809 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH8.0), 10% glycerol |
| Gene ID (Entrez) | 55312 |
| Molecular Weight (g/mol) | 20.5kDa |
| Name | RFK Protein |
| Purification Method | Protein |
| Quantity | 0.1 mg |
| Immunogen | MGSSHHHHHH SSGLVPRGSH MPRADCIMRH LPYFCRGQVV RGFGRGSKQL GIPTANFPEQ VVDNLPADIS TGIYYGWASV GSGDVHKMVV SIGWNPYYKN TKKSMETHIM HTFKEDFYGE ILNVAIVGYL RPEKNFDSLE SLISAIQGDI EEAKKRLELP EHLKIKEDNF FQVSKSKIMN GH |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
