Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Separase Antibody Blocking Peptide

Click to view available options
Quantity:
0.05 mg
Description
A peptide to ESPL1.
Specifications
Specifications
Gene ID (Entrez) | 9700 |
Species | Human |
Content And Storage | Store at -80 C. Avoid freeze-thaw cycles. |
Formulation | Peptide dissolved in dH2O. Contains no BSA. |
For Use With (Application) | This Peptide is Useful as a Blocking Peptide for BC110-55295. For Further Blocking Peptide Related Protocol, Click Here . |
Gene Alias | Caspase-like protein ESPL1, EC 3.4.22.49, ESP1extra spindle poles like 1, extra spindle pole bodies homolog 1 (S. cerevisiae), extra spindle poles like 1 (S. cerevisiae), Extra spindle poles-like 1 protein, FLJ46492, KIAA0165separin, Separase |
Gene Symbol | ESPL1 |
Product Type | Separase |
Quantity | 0.05 mg |
Regulatory Status | RUO - research use only |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction