Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ SLC39A13 Antibody Blocking Peptide
SDP

Catalog No. NBP176509E Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176509E 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176509E Supplier Novus Biologicals™ Supplier No. NBP176509PEP
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A blocking peptide containing 14 amino acid peptide near the amino terminus of human ZIP13. Source: Synthetic (Accession #: NP_001121697) The SLC39A13 Blocking Peptide has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 91252
Species Human
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-76509. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias FLJ25785, LIV-1 subfamily of ZIP zinc transporter 9, LZT-Hs9, solute carrier family 39 (metal ion transporter), member 13, solute carrier family 39 (zinc transporter), member 13, Solute carrier family 39 member 13, zinc transporter ZIP13, ZIP13, ZIP-13, Zrt- and Irt-like protein 13
Gene Symbol SLC39A13
Product Type SLC39A13
Quantity 0.05 mg
Regulatory Status RUO - research use only
Source Synthetic
Termination 14 amino acid peptide near the amino terminus of human ZIP13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.