Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ SOD1/Cu-Zn SOD Protein
SDP

Catalog No. NBP17229001 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17229001 0.1 mg
NBP1722905 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17229001 Supplier Novus Biologicals™ Supplier No. NBP1722900.1MG
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-206 of E.coli SOD1/Cu-Zn SOD The Recombinant E. coli SOD1/Cu-Zn SOD Protein is derived from E. coli. The Recombinant E. coli SOD1/Cu-Zn SOD Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 0.1M NaCl
Molecular Weight (g/mol) 25.2kDa
Purification Method SDS-PAGE
Quantity 0.1 mg
Source Bacteria
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name SOD1/Cu-Zn SOD
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.