Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ SPOP Protein Carrier Free

Click to view available options
Quantity:
0.05 mg
0.25 mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-374 of Human SPOP The Recombinant Human SPOP Protein is derived from E. coli. The Recombinant Human SPOP Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Concentration | 0.5mg/mL |
For Use With (Application) | SDS-PAGE |
Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 5mM DTT, 50% glycerol, 0.2M NaCl, 2mM EDTA |
Molecular Weight (g/mol) | 44.3kDa |
Purification Method | SDS-PAGE |
Quantity | 0.05 mg |
Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
Regulatory Status | RUO |
Common Name | SPOP |
Conjugate | Unconjugated |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction