Learn More
Specifications
Specifications
| Gene ID (Entrez) | 54434 |
| Purification Method | Affinity purified |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | Positive Control |
| Product Type | SSH1 Peptide |
| Quantity | 0.25 mg |
| Regulatory Status | RUO - research use only |
| Termination | beta Amyloid aa 3-42 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
