Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TMEM97 Antibody Blocking Peptide
SDP

Catalog No. NBP130436E Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130436E 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130436E Supplier Novus Biologicals™ Supplier No. NBP130436PEP
Add to Cart
Add to Cart
A peptide to TMEM97.

Specifications

Gene ID (Entrez) 27346
Species Human
Content And Storage Store at -80 C. Avoid freeze-thaw cycles.
Formulation Peptide dissolved in dH2O. Contains no BSA.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NBP1-30436. For Further Blocking Peptide Related Protocol, Click Here .
Gene Alias MAC30Protein MAC30, transmembrane protein 97
Gene Symbol TMEM97
Product Type TMEM97
Quantity 0.05 mL
Regulatory Status RUO - research use only
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.