Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Triosephosphate isomerase Protein
SDP

Catalog No. NBP14945101
Click to view available options
Quantity:
0.1 mg
0.5 mg

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-249 of Human Triosephosphate isomerase The Recombinant Human Triosephosphate isomerase Protein is derived from E. coli. The Recombinant Human Triosephosphate isomerase Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_000356
Concentration 0.5mg/mL
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol, 1mM DTT
Gene ID (Entrez) 7167
Molecular Weight (g/mol) 31kDa
Purification Method Protein
Quantity 0.1 mg
Source Recombinant human TPI1 protein, fused to His-tag at N-terminus, expressed in E. coli
Immunogen TPI1, 1-249aa, Human, His tag, E. coli (NP_000356).
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Common Name Triosephosphate isomerase
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.