Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ TSTD1 Protein

Click to view available options
Quantity:
250 μg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-115 of Human TSTD1 The Recombinant Human TSTD1 Protein is derived from E. coli. The Recombinant Human TSTD1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Concentration | 1mg/mL |
For Use With (Application) | SDS-PAGE |
Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol,100mM NaCl |
Quantity | 250 μg |
Source | Human |
Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Regulatory Status | RUO |
Common Name | TSTD1 |
Conjugate | Unconjugated |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction