Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ v6.5 Mouse embryonic stem cells Whole Cell Lysate
Shop All Novus Biologicals Products

Click to view available options
Quantity:
0.25 mg
Description
The v6.5 Mouse embryonic stem cells Whole Cell Lysate is derived from mouse. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Tissue | Embryo |
For Use With (Application) | Immunoprecipitation, Western Blot |
Quantity | 0.25 mg |
Storage Buffer | Standard RIPA buffer including protease and phosphatase inhibitors and 0.1% SDS. No reducing agents or loading buffer added. |
Purity | Protein |
Cellular Fraction | Whole |
Content And Storage | -80°C. Avoid freeze-thaw cycles. |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction