Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ v6.5 Mouse Embryonic Stem Cells
SDP

Catalog No. NBP141162 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP141162 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP141162 Supplier Novus Biologicals™ Supplier No. NBP141162

Applications: In vitro assay

  • Cells are supplied in 2mL quantities (about 1 x 106 cells/mL) in freezing media (60% DMEM, 20% FBS, 20% DMSO)
Applications: In vitro assay

Recommended Storage

Store in gas phase of liquid nitrogen

Warranty and Services

6 Months

Specifications

Content And Storage Store in gas phase of liquid nitrogen
For Use With (Application) In vitro assay
Quantity 2mL
Source Male mouse embryo

This product is for research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.