Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant E. coli cysH His Protein
SDP

Catalog No. NBP222650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222650 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222650 Supplier Novus Biologicals™ Supplier No. NBP222650
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-244 of E.coli cysH The Recombinant E. coli cysH Protein is derived from E. coli. The Recombinant E. coli cysH Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4), 1mM DTT, 20% glycerol
Gene ID (Entrez) 947230
Molecular Weight (g/mol) 30.1kDa
Name cysH Protein
Purification Method SDS-PAGE
Quantity 0.1 mg
Source E. Coli
Immunogen cysH, 1-244aa, E.coli, His tag, E.coli MGSSHHHHHH SSGLVPRGSH MSKLDLNALN ELPKVDRILA LAETNAELEK LDAEGRVAWA LDNLPGEYVL SSSFGIQAAV SLHLVNQIRP DIPVILTDTG YLFPETYRFI DELTDKLKLN LKVYRATESA AWQEARYGKL WEQGVEGIEK YNDINKVEPM NRALKELNAQ TWFAGLRREQ SGSRANLPVL AIQRGVFKVL PIIDWDNRTI YQYLQKHGLK YHPLWDEGYL SVGDTHTTRK WEPGMAEEET RFFGLKRECG LHEG
Storage Requirements −20°C or −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.