Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Yeast Hsp104 Protein
SDP

Catalog No. NBC118363 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118363 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118363 Supplier Novus Biologicals™ Supplier No. NBC118363
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 1-908 of Saccharomyces cerevisiae Hsp104 The Recombinant Yeast Hsp104 Protein is derived from E. coli. The Recombinant Yeast Hsp104 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_013074
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer, (pH 8.0), 100mM NaCl, 2mM EDTA, 5% glycerol
Gene ID (Entrez) 850633
Molecular Weight (g/mol) 102kDa
Name Hsp104 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen Hsp104(amino acids 1-908) Saccharomyces cerevisiae, Recombinant, E.coli (NP_013074).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Yeast
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.