Learn More
Tris-EDTA Buffer Solutions, Novus Biologicals™
SDP
For recovery of antigens masked by fixation in cross-linking fixatives, such as formalin
$280.00 - $452.00
Specifications
Quantity 500 mL
View More Specs
Products 2
Catalog Number Mfr. No. pH Price Quantity & Availability  
Catalog Number Mfr. No. pH Price Quantity & Availability  
NB90062085
SDP
View Documents
Novus Biologicals
NB90062085
9
Each for $452.00
Only null left
Add to Cart
NB90066730
SDP
View Documents
Novus Biologicals
NB90066730
8
Each for $280.00
Only null left
Add to Cart
Description
Description
  • 10X EDTA buffer for Immunohistochemistry
  • Dilute one part buffer with nine parts distilled water
  • Unit size: 500mL
Specifications
Specifications
500 mL
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.