Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597106
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597106 Supplier Invitrogen™ Supplier No. PA597106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSIWYKFQCA DHNLKTKKIY FYWICRETGA FSWFNNLLTS LEQEMEELGK VGFLNYRLFL TGWDSNIVGH AALNFDKATD IVTGLKQKTS FGRPMWDNEF STIATSHPKS VVGVFLCGPR TLAKSLRKCC HRYSSLDPRK VQFYFNKENF; Positive Samples: HT-29, A-549, SW480; Cellular Location: Cell projection, Multi-pass membrane protein, invadopodium membrane.

Voltage-gated proton (hydrogen) channels play an important role in cellular defense against acidic stress. NOX1 is a homolog of the catalytic subunit of the superoxide-generating NADPH oxidase of phagocytes, gp91phox. Three splice variants of NOX1 have been identified, NOH-1L, NOH-1S and NOH-1Lv. NOH-1S is a voltage-gated proton channel that participates in the regulation of cellular pH and is blocked by zinc. NOH-1L is a pyridine nucleotide-dependent oxidoreductase that generates superoxide and might conduct H (+) ions as part of its electron transport mechanism, whereas NOH-1S does not contain an electron transport chain. NOX1 have the potential to be effective treatments for a range of ischemic diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOX1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Nox1
Gene Accession No. Q8CIZ9, Q9WV87, Q9Y5S8
Gene Alias GP91 phox homolog; GP91-2; mitogenic oxidase (pyridine nucleotide-dependent superoxide-generating); mitogenic oxidase 1; MOX1; MOX-1; NADH/NADPH mitogenic oxidase subunit P65-MOX; NADPH oxidase 1; NADPH oxidase 1 alpha; NADPH oxidase homolog-1; NOH1; NOH-1; Nox1; NOX-1; NOX1a; NOX1alpha
Gene Symbols Nox1
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114243, 237038, 27035
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.