Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOX1 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142645
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_008983.2; NP_39249.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
Voltage-gated proton (hydrogen) channels play an important role in cellular defense against acidic stress. NOX1 is a homolog of the catalytic subunit of the superoxide-generating NADPH oxidase of phagocytes, gp91phox. Three splice variants of NOX1 have been identified, NOH-1L, NOH-1S and NOH-1Lv. NOH-1S is a voltage-gated proton channel that participates in the regulation of cellular pH and is blocked by zinc. NOH-1L is a pyridine nucleotide-dependent oxidoreductase that generates superoxide and might conduct H (+) ions as part of its electron transport mechanism, whereas NOH-1S does not contain an electron transport chain. NOX1 have the potential to be effective treatments for a range of ischemic diseases.
Specifications
| NOX1 | |
| Polyclonal | |
| Unconjugated | |
| Nox1 | |
| GP91 phox homolog; GP91-2; mitogenic oxidase (pyridine nucleotide-dependent superoxide-generating); mitogenic oxidase 1; MOX1; MOX-1; NADH/NADPH mitogenic oxidase subunit P65-MOX; NADPH oxidase 1; NADPH oxidase 1 alpha; NADPH oxidase homolog-1; NOH1; NOH-1; Nox1; NOX-1; NOX1a; NOX1alpha | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 27035 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9Y5S8 | |
| Nox1 | |
| Peptide with sequence C-DKATDIVTGLKQK (aa 490-502). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction