Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOX2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577938
Description
Positive Control: 293T, HeLa, HepG2 Predicted Reactivity: Rabbit (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Mox1 and the glycoprotein gp91-phox are largely related proteins that are essential components of the NADPH oxidase. The superoxide-generating NADPH oxidase complex expresses in phagocytes, neuroepithelial bodies, vascular smooth muscle cells, and endothelial cells. It includes a membrane bound flavocytochrome containing two subunits, gp91-phox and p22-phox, and the cytosolic proteins p47-phox and p67-phox. The p22- and gp91-phox subunits also function as surface O2 sensors that initiate cellular signaling in response to hypoxic conditions. Mox1 and gp91 contain identical C-terminal sequence identity, yet possess distinct expression patterns. gp91-phox expresses in eosinophils, neutrophils, monocytes, and B-lymphocytes, whereas Mox1 is predominantly detected in the colon, with low expression in the uterus and prostate.
Specifications
NOX2 | |
Polyclonal | |
Unconjugated | |
Cybb | |
AMCBX2; C88302; Cgd; CGD91-phox; Cybb; Cyd; cytochrome b(558) subunit beta; cytochrome b-245 beta chain; cytochrome b-245 beta polypeptide; cytochrome b-245 heavy chain; cytochrome b-245, beta polypeptide; cytochrome b558 subunit beta; endothelial type gp91-phox; GP91 PHOX; GP911; GP91-1; GP91PHOX; gp91-phox; Heme-binding membrane glycoprotein gp91phox; IMD34; NADPH oxidase 2; neutrophil cytochrome b 91 kDa polypeptide; Nox2; p22 phagocyte B-cytochrome; p91PHOX; p91-PHOX; superoxide-generating NADPH oxidase heavy chain subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13058, 1536 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.73 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P04839, Q61093 | |
Cybb | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Human NOX2/gp91phox. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction