Learn More
Description
Specifications
Specifications
| Antigen | Nox4 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, KnockDown |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature, Immunoblotting reported in scientific literature (PMID 28152080), KnockDown Validated |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | Q9NPH5 |
| Gene Alias | EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOXEC 1.6.3.- |
| Gene Symbols | NOX4 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
