Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nox4 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP241293
Description
Nox4 Polyclonal specifically detects Nox4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
Nox4 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
NOX4 | |
Antibody was raised against a 14 amino acid peptide near the amino terminus of human NOX4. The immunogen is located within amino acids 70 - 120 of NOX4. Amino Acid Squence: RGSQKVPSRRTRRL | |
Affinity Purified | |
RUO | |
Primary | |
At least four isoforms of NOX4 are known to exist. NOX4 is predicted to not cross-react with other NOX proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL, Immunofluorescence 20 μg/mL | |
EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOXEC 1.6.3.- | |
Rabbit | |
68 kDa | |
0.1 mg | |
Cancer, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Signal Transduction | |
50507 | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction