Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nox4 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11058851MFV500
Description
Nox4 Polyclonal antibody specifically detects Nox4 in Human, Mouse, Rat, Porcine, Primate samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
Nox4 | |
Polyclonal | |
Flow Cytometry, Flow (Intracellular) | |
EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOXEC 1.6.3.- | |
A synthetic peptide made to a C-terminal region (within residues 500-578) of the human NOX4 protein sequence. [Swiss-Prot# Q9NPH5]. | |
0.1 mL | |
Cancer, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Signal Transduction | |
50507 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction