Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nox4 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299109MFV610
Description
Nox4 Polyclonal antibody specifically detects Nox4 in Human samples. It is validated for Western BlotSpecifications
Nox4 | |
Polyclonal | |
Western Blot | |
EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOXEC 1.6.3.- | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Nox4. | |
0.1 mL | |
Cancer, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Signal Transduction | |
50507 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction