Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOX4 Mouse anti-Human, Rat, Clone: 4E5F1, ProteinTech
SDP

Catalog No. 502239740 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-9740 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-9740 Supplier Proteintech Group Inc Supplier No. 676811IG150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Aliquoting is unnecessary for -20°C storage.

NOX4 Monoclonal antibody specifically detects NOX4 in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen NOX4
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4E5F1
Concentration 1000 μg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Nox4
Gene Accession No. Q924V1, Q9NPH5
Gene Alias AI648021; kidney oxidase-1; Kidney superoxide-producing NADPH oxidase; KOX; KOX1; KOX-1; NADPH oxidase 4; NOX 4; NOX4; NOX-4; renal NAD(P)H-oxidase; RENOX; superoxide-generating NADPH oxidase 4
Gene Symbols Nox4
Host Species Mouse
Immunogen NOX4 Fusion Protein Ag6176
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50507, 85431
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.