Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ NOX4, Mouse, Unlabeled, Clone: 10A8.1,

Catalog No. MABC616MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABC616MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC616MI Supplier MilliporeSigma™ Supplier No. MABC616
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NADPH oxidase 4 (UniProt Q9NPH5; also known as Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX-1, Renal NAD(P)H-oxidase) is encoded by the NOX4 (also known as RENOX) gene (Gene ID 50507) in human. NOX4 belongs to the family of NADPH oxidases that modulate redox-dependent cellular signalling pathways via intracellular reactive oxygen species (ROS) generation. Seven family members have been identified in mammals (NOX1–5 and DUOX1/2) with member-specific cellular distribution, intracellular location, mechanisms of action and types of ROS produced. NOX4 produces predominantly H2O2, and is widely expressed in many cell types and adult tissues, including kidney, liver, heart and aorta. NOX4 plays an important role in homocysteine (Hcy) metabolism, which in turns is a determining factor that impacts on defence against environmental toxins. Nox-knocout in mice results in significantly decreased total plasma Hcy levels, causing markedly increased susceptibility to acetaminophen- (APAP-, paracetamol-) induced hepatoxicity as a result of significantly decreased cysteine production and hepatic GSH stores. Human NOX4 is a six-transmembrane (a.a. 17-37, 63-83, 104-124, 155-175, 189-209, 425-445) protein, having both its N- and C-terminal ends at the cytoplasmic side (a.a. 1-16 and 446-578).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOX4
Applications Western Blot
Classification Monoclonal
Clone 10A8.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Mouse monoclonal IgG2bκ hybridoma culture supernatant without preservatives.
Gene NOX4, RENOX
Gene Accession No. Q9NPH5
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to a fragment from the third extracellular loop of human NOX4.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001137308
Test Specificity Immunogen construct region is present in human NOX4 spliced isoform 1, 2 (Nox4A), and 8, missing in isoform 5 (Nox4C), 7, and 9, partially truncated in isoform 3 (Nox4E), 4 (Nox4D), and 6 (Nox4B) reported by UniProt (Q9NPH5).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Culture Supernatant
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.