Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOX4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585479
Description
Keep as concentrated solution. Predicted reactivity: Mouse (93%), Rat (93%), Bovine (96%). Positive Control: 293T, HeLa, A431, Jurkat, Raji, unboiled NOX4-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Oxygen sensing is essential for homeostasis in all aerobic organisms. A phagocyte-type oxidase, similar to that responsible for the production of large amounts of reactive oxygen species (ROS) in neutrophil granulocytes, with resultant antimicrobial activity, has been postulated to function in the kidney as an oxygen sensor that regulates the synthesis of erythropoietin in the renal cortex. NOX4 has a role as a redox messenger in the activation of intracellular signaling pathways leading (or contributing) to mitochondriogenesis, cell survival, and differentiation in hematopoietic stem cells. Data suggests that NOX4 provides a novel link between the insulin receptor and the generation of cellular reactive oxygen species that enhances insulin signal transduction.
Specifications
NOX4 | |
Polyclonal | |
Unconjugated | |
Nox4 | |
AI648021; kidney oxidase-1; Kidney superoxide-producing NADPH oxidase; KOX; KOX1; KOX-1; NADPH oxidase 4; NOX 4; NOX4; NOX-4; renal NAD(P)H-oxidase; RENOX; superoxide-generating NADPH oxidase 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50490, 50507, 85431 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
1.84 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q924V1, Q9JHI8, Q9NPH5 | |
Nox4 | |
Recombinant protein encompassing a sequence within the center region of human NOX4. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction