Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOX4 Recombinant Rabbit Monoclonal Antibody (2B2K7)
GREENER_CHOICE

Catalog No. FEMA551408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551408 Supplier Invitrogen™ Supplier No. MA551408
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: LHNKFWQENR PDYVNIQLYL SQTDGIQKII GEKYHALNSR LFIGRPRWKL LFDEIAKYNR GKTVGVFCCG PNSLSKTLHK LSNQNNSYGT RFEYNKESFS.

Oxygen sensing is essential for homeostasis in all aerobic organisms. A phagocyte-type oxidase, similar to that responsible for the production of large amounts of reactive oxygen species (ROS) in neutrophil granulocytes, with resultant antimicrobial activity, has been postulated to function in the kidney as an oxygen sensor that regulates the synthesis of erythropoietin in the renal cortex. NOX4 has a role as a redox messenger in the activation of intracellular signaling pathways leading (or contributing) to mitochondriogenesis, cell survival, and differentiation in hematopoietic stem cells. Data suggests that NOX4 provides a novel link between the insulin receptor and the generation of cellular reactive oxygen species that enhances insulin signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOX4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 2B2K7
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Nox4
Gene Accession No. Q924V1, Q9JHI8, Q9NPH5
Gene Alias AI648021; kidney oxidase-1; Kidney superoxide-producing NADPH oxidase; KOX; KOX1; KOX-1; NADPH oxidase 4; NOX 4; NOX4; NOX-4; renal NAD(P)H-oxidase; RENOX; superoxide-generating NADPH oxidase 4
Gene Symbols Nox4
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50490, 50507, 85431
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.