Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOX5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596588 Supplier Invitrogen™ Supplier No. PA596588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: 22Rv1; Cellular Location: Endoplasmic reticulum, Membrane, Multi-pass membrane protein Immunogen sequence: AEEAQYGRFL ELHMYMTSAL GKNDMKAIGL QMALDLLANK EKKDSITGLQ TRTQPGRPDW SKVFQKVAAE KKGKVQVFFC GSPALAKVLK GHCEKFGFRF FQENF.

This gene is predominantly expressed in the testis and lymphocyte-rich areas of spleen and lymph nodes. It encodes a calcium-dependen NADPH oxidase that generates superoxide, and functions as a calcium-dependent proton channel that may regulate redox-dependent processes in lymphocytes and spermatozoa. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq, Oct 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOX5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene NOX5
Gene Accession No. Q96PH1
Gene Alias NADPH oxidase 5; NADPH oxidase, EF-hand calcium binding domain 5; NOX5; NOX5zeta
Gene Symbols NOX5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 661-765 of human NOX5 (NP_0787813).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79400
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.