Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Noxa Antibody (114C307.1) - Non-Recombinant Monoclonal, Novus Biologicals™
SDP

Catalog No. NB6001159SS Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001159SS 0.025 mg
NB6001159 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB6001159SS Supplier R&D Systems Supplier No. NB6001159SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Noxa Monoclonal specifically detects Noxa in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Noxa
Applications Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 114C307.1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1 ug/ million cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:20-1:1000, Immunohistochemistry-Paraffin 5 ug/ml
Gene Accession No. Q13794
Gene Alias adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa
Gene Symbols PMAIP1
Host Species Mouse
Immunogen This Noxa antibody was developed by immunizing mice with a fusion protein containing human Noxa.
Molecular Weight of Antigen 14.96 kDa
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Membrane Trafficking and Chaperones, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5366
Test Specificity An ∼80 kD background band may also be seen in Jurkat cell lysate.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.