Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Noxa Antibody (69N15C9), Alexa Fluor™ 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP225266AF350
Description
Noxa Monoclonal specifically detects Noxa in Mouse samples. It is validated for Western Blot.Specifications
Noxa | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
PMAIP1 | |
This Noxa antibody was developed by immunizing mice with a fusion protein containing human Noxa. | |
Protein G purified | |
Apoptosis, Cancer, Cellular Markers, Membrane Trafficking and Chaperones, Necroptosis, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors | |
5366.0 | |
Store at 4°C in the dark. |
Western Blot | |
69N15C9 | |
Western Blot | |
adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa | |
Rat | |
11.6 kDa | |
0.1 mL | |
Primary | |
Mouse | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction