Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOXA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112629
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112629 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112629 Supplier Invitrogen™ Supplier No. PA5112629
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NOXA1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

NADPH oxidases (NOXs) catalyze the transfer of electrons from NADPH to molecular oxygen to generate reactive oxygen species (ROS). The NOX activator NOXA1 can stimulate both NOX1 and NOX2, but it appears to be more effective in activating NOX1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOXA1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NOXA1
Gene Accession No. Q86UR1, Q8CJ00
Gene Alias antigen NY-CO-31; inhibitory NADPH oxidase activator 1; NADPH oxidase activator 1; NCF2-like protein; Nox activator 1; Noxa1; NY-CO-31; p51NOX; p51-nox; P67phox-like factor; predicted NADPH oxidase activator 1; SDCCAG31; serologically defined colon cancer antigen 31
Gene Symbols NOXA1
Host Species Rabbit
Immunogen Recombinant Human NADPH oxidase activator 1 protein (150-313AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10811, 241275
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.