Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOXA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526572
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526572 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526572 Supplier Thermo Scientific Supplier No. PA526572
Add to Cart
Add to Cart

NOXA1 Polyclonal Antibody

NOXA1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen NOXA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene NOXA1
Gene Accession No. Q86UR1
Gene Alias antigen NY-CO-31; inhibitory NADPH oxidase activator 1; NADPH oxidase activator 1; NCF2-like protein; Nox activator 1; Noxa1; NY-CO-31; p51NOX; p51-nox; P67phox-like factor; predicted NADPH oxidase activator 1; SDCCAG31; serologically defined colon cancer antigen 31
Gene Symbols NOXA1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 449-478 amino acids from the C-terminal region of human NOXA1.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10811
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.