Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOXA1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS19322R

Description
NADPH oxidases (NOXs) catalyze the transfer of electrons from NADPH to molecular oxygen to generate reactive oxygen species (ROS). The NOX activator NOXA1 can stimulate both NOX1 and NOX2, but it appears to be more effective in activating NOX1.Specifications
NOXA1 | |
Polyclonal | |
Unconjugated | |
NOXA1 | |
Antigen NY-CO-31; inhibitory NADPH oxidase activator 1; NADPH oxidase activator 1; NCF2-like protein; NOX activator 1; NOXA1; NY-CO-31; P51NOX; p51-nox; P67phox-like factor; SDCCAG31; serologically defined colon cancer antigen 31 | |
Rabbit | |
Protein A | |
RUO | |
10811 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q86UR1 | |
NOXA1 | |
KLH conjugated synthetic peptide derived from human NOXA1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction