Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOXO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564466
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564466 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564466 Supplier Invitrogen™ Supplier No. PA564466
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLETYSRRLL ATAERVARSP TITGFFAPQP LDLE Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 62%.

NADPH oxidases catalyze the transfer of electrons from NADPH to molecular oxygen to generate reactive oxygen species to NOX and also target NOX to different subcellular compartments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOXO1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NOXO1
Gene Accession No. Q8NFA2
Gene Alias 2310034C04Rik; hslt; MGC20258; NADPH oxidase organizer 1; NADPH oxidase regulatory protein; Nox organizer 1; NOXO1; Nox-organizing protein 1; P41NOX; P41NOXA; P41NOXB; P41NOXC; regulatory protein P41NOX; SH3 and PX domain-containing protein 5; SH3PXD5; Snx28; Sorting nexin-28
Gene Symbols NOXO1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NOXO1. Recombinant protein control fragment (Product #RP-103911).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124056
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.