Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOXO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110038 Supplier Invitrogen™ Supplier No. PA5110038
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QPLDLEPALP PGSRVILPTP EEQPLSRAAG RLSIHSLEAQ SLRCLQPFCT QDTRDRPFQA QAQESLDVLL RHPSGWWLVE NEDRQTAWFP APYLEEAAPG QGREGGPSLG SSGPQFCASR AYESSRADEL SVPAGARVRV LETSDRGWWL CRYGDRAGLL PAVLLRPEGL GALLSGTGFR GGDDPAGEAR GFPEPSQATA PPPTVPTRPS PGAIQSRCCT VTRRALERRP RRQGRPRGCV DSVPHPTTEQ.

NADPH oxidases catalyze the transfer of electrons from NADPH to molecular oxygen to generate reactive oxygen species to NOX and also target NOX to different subcellular compartments.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOXO1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NOXO1
Gene Accession No. Q8VCM2
Gene Alias 2310034C04Rik; hslt; MGC20258; NADPH oxidase organizer 1; NADPH oxidase regulatory protein; Nox organizer 1; NOXO1; Nox-organizing protein 1; P41NOX; P41NOXA; P41NOXB; P41NOXC; regulatory protein P41NOX; SH3 and PX domain-containing protein 5; SH3PXD5; Snx28; Sorting nexin-28
Gene Symbols NOXO1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 121-370 of human NOXO1 (NP_653204.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71893
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.