Learn More
Invitrogen™ NOXO1 Polyclonal Antibody

Description
Immunogen sequence: HSLEAQSLRC LQPFCTQDTR DRPFQAQAQE SLDVLLRHPS GWWLVENEDR For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 77%; Horse: 77%; Human: 100%.
Specifications
Specifications
| Antigen | NOXO1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | NOXO1 |
| Gene Accession No. | Q8NFA2 |
| Gene Alias | 2310034C04Rik; hslt; MGC20258; NADPH oxidase organizer 1; NADPH oxidase regulatory protein; Nox organizer 1; NOXO1; Nox-organizing protein 1; P41NOX; P41NOXA; P41NOXB; P41NOXC; regulatory protein P41NOX; SH3 and PX domain-containing protein 5; SH3PXD5; Snx28; Sorting nexin-28 |
| Gene Symbols | NOXO1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.