Learn More
Invitrogen™ NOXO1 Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000. Preliminary analysis showed no signal in Western Blot.
Specifications
Specifications
| Antigen | NOXO1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | NOXO1 |
| Gene Accession No. | Q8NFA2 |
| Gene Alias | 2310034C04Rik; hslt; MGC20258; NADPH oxidase organizer 1; NADPH oxidase regulatory protein; Nox organizer 1; NOXO1; Nox-organizing protein 1; P41NOX; P41NOXA; P41NOXB; P41NOXC; regulatory protein P41NOX; SH3 and PX domain-containing protein 5; SH3PXD5; Snx28; Sorting nexin-28 |
| Gene Symbols | NOXO1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.