Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOXRED1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569855
Description
This target displays homology in the following species: Cow: 85%; Dog: 100%; Guinea Pig: 77%; Human: 100%; Mouse: 85%.
Probable oxidoreductase.
Specifications
NOXRED1 | |
Polyclonal | |
Unconjugated | |
NOXRED1 | |
4933437F05Rik; C14orf148; NADP dependent oxidoreductase domain containing 1; NADP+ dependent oxidoreductase domain containing 1; NADP-dependent oxidoreductase domain containing 1; NADP-dependent oxidoreductase domain-containing protein 1; Noxred1; Pyrroline-5-carboxylate reductase-like protein C14orf148; pyrroline-5-carboxylate reductase-like protein C14orf148 homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
122945 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6NXP6 | |
NOXRED1 | |
synthetic peptide directed towards the middle region of human NOXRED1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction