Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOXRED1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563155 Supplier Invitrogen™ Supplier No. PA563155
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILNIKWLEGV FYAALNICTA RNMAHSQVLQ LLSELFLSVH FEDCGKDTAS CPKLQLTDFV SKAYGKNLSQ ERPFPWFDLT AVQLKETPFS QHLSSSPVLQ DHL Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 66%.

Probable oxidoreductase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOXRED1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NOXRED1
Gene Accession No. Q6NXP6
Gene Alias 4933437F05Rik; C14orf148; NADP dependent oxidoreductase domain containing 1; NADP+ dependent oxidoreductase domain containing 1; NADP-dependent oxidoreductase domain containing 1; NADP-dependent oxidoreductase domain-containing protein 1; Noxred1; Pyrroline-5-carboxylate reductase-like protein C14orf148; pyrroline-5-carboxylate reductase-like protein C14orf148 homolog
Gene Symbols NOXRED1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NOXRED1. Recombinant protein control fragment (Product #RP-99869).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122945
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.