Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ NP-40 Alternative, Calbiochem™,

Catalog No. 4920161000M
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Packaging:
Glass Bottle

Chemical Identifiers

CAS 9016-45-9
Molecular Formula C19H32O3
Molecular Weight (g/mol) 308.46
MDL Number MFCD00134080
InChI Key BLXVTZPGEOGTGG-UHFFFAOYSA-N
Synonym Nonylphenyl Polyethylene Glycol
PubChem CID 24773
IUPAC Name 2-[2-(4-nonylphenoxy)ethoxy]ethan-1-ol
SMILES CCCCCCCCCC1=CC=C(OCCOCCO)C=C1

Specifications

Color Undesignated
Quantity 1000 mL
Packaging Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.